Learn More
Empore™ Sealing Tape for 96-well Plate, Sold by MilliporeSigma™ Supelco™
Catalog No. 111022331 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Polypropylene Filter (graded density)
Catalog No. Material
11-102-2331 Polypropylene Filter (graded density)
1 options
Catalog No. 11-102-2331 Supplier MilliporeSigma Supelco Supplier No. 66881U
Add to Cart
Add to Cart
Designed for sealing unused portions of a 96-well filter/SPE plate during vacuum processing. Each sheet is comprised of a polypropylene fil coated with a synthetic rubber adhesive and is coupled with a liner tab for easy handling and dispensing
  • Comprises of SPE particles tightly enmeshed within a network of inert PTFE fibrils (90% SPE particles : 10% PTFE, by weight).
  • Reduced elution volume (≤ 100 μL) allows for direct inject procedure or reduced eluate evaporation times
  • Allows for faster flow-rates without risk of recovery and reproducibility loss
  • Proprietary pre-filter reduces risk of clogging
  • Luer tip collar eliminates potential cross-contamination
TRUSTED_SUSTAINABILITY
Specifications
Bed Volume (Metric) 18 μL
Volume (Metric) Well 1.2 mL
Material Polypropylene Filter (graded density)
Quantity 10/Pk.
Diameter (Metric) Inner 5.5 mm
Empore is a trademark of CDS Analytical