Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Empore™ SPE 96-Well Plate, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022328 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Particle Size:
32 μm
50 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
11-102-2328 50 μm
11-102-2329 32 μm
11-102-2330 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2328 Supplier MilliporeSigma Supelco Supplier No. 66875U
Add to Cart
Add to Cart

Comprises of SPE particles tightly enmeshed within a network of inert PTFE fibrils (90% SPE particles : 10% PTFE, by weight).

  • Results in a highly dense and uniform extraction medium that offers distinct advantages over traditional sorbent/packed-bed SPE products.
  • Provides a denser, more uniform extraction bed than traditional packed bed products allowing for smaller bed weights, shorter analyte to pore diffusion paths, and more efficient extractions.
  • Ideal for high throughput SPE allowing users to process up to 96 samples in parallel.
  • The PP pre-filter acts as depth filter that provides faster flow-rates and reduces the risk of clogging.
  • Reduced elution volume (≤ 100 μL) allows for direct inject procedure or reduced eluate evaporation times
  • Allows for faster flow-rates without risk of recovery and reproducibility loss
  • Recommended for the high-throughput extraction of moderately-polar to non-polar analytes from aqueous samples (e.g., biological plasma).
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume (Metric) 18 μL
Bed Weight 10 to 15 mg/well
Particle Size 50 μm
Type C18
Volume (Metric) Well 1.2 mL
Endcapped Encapped
Material Polypropylene filter (graded density)
Pore Size 60 Å
Quantity 1/Pk.
Diameter (Metric) Inner 5.5 mm

Empore is a trademark of CDS Analytical

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.