Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Empore™ SPE 96-Well Plate, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022330 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Particle Size:
32 μm
50 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
11-102-2330 -
11-102-2328 50 μm
11-102-2329 32 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2330 Supplier MilliporeSigma Supelco Supplier No. 66878U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Comprises of SPE particles tightly enmeshed within a network of inert PTFE fibrils (90% SPE particles : 10% PTFE, by weight).

  • The plate provides an excellent media for trapping visible particulates from samples in a 96-well format, while maintaining excellent flow characteristics.
  • Ideal for removing denatured protein particulates from biological plasma after protein precipitation.
  • Ideal for high throughput SPE allowing users to process up to 96 samples in parallel.
  • The unique Empore technology comprises of a series of polypropylene (PP) pre-filters that are layed on top of the SPE disk.
  • The PP pre-filter acts as depth filter that provides faster flow-rates and reduces the risk of clogging.
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume (Metric) 18 μL
Volume (Metric) Well 1.2 mL
Material Polypropylene filter (graded density)
Pore Size ≥10 μm retention, 98% retention
Quantity 1/Pk.
Diameter (Metric) Inner 5.5 mm

Empore is a trademark of CDS Analytical

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.