Learn More
Thomas Scientific Tubes (empty) pack of 1000 with caps and sealing ring
SDP
Catalog No. 501461318
Encompass_Preferred
Supplied in bulk or in 2.0mL tubes-prefilled with beads Compatible with all common bead homogenizers Available in glass, ceramic or stainless steel Unique Triple-Pure™ grade: Acid washed, heat treated and tested free of nuclease Bead beating is quickly becoming the preferred process used to break up (lyse) samples in order to release the DNA, RNA and proteins contained within cells. Samples are placed in tubes with the appropriate grinding beads and subjected to high energy mixing. The beads impact the sample, eventually breaking it down on the cellular level releasing subcellular contents. A variety of prefilled tubes are available for homogenizing samples including those with stainless steel, glass and Triple-Pure™ zirconium beads. All three bead types are acid washed and processed to remove nucleases and proteases. The Triple-Pure bead tubes undergo special handling during filling and are quality tested to ensure the elimination of DNase, RNase, protease, and nucleic aci
Catalog No. 50-146-1318 Supplier Thomas Scientific Supplier No. 1207X15
Supplied in bulk or in 2.0mL tubes-prefilled with beads Compatible with all common bead homogenizers Available in glass, ceramic or stainless steel Unique Triple-Pure™ grade: Acid washed, heat treated and tested free of nuclease Bead beating is quickly becoming the preferred process used to break up (lyse) samples in order to release the DNA, RNA and proteins contained within cells. Samples are placed in tubes with the appropriate grinding beads and subjected to high energy mixing. The beads impact the sample, eventually breaking it down on the cellular level releasing subcellular contents. A variety of prefilled tubes are available for homogenizing samples including those with stainless steel, glass and Triple-Pure™ zirconium beads. All three bead types are acid washed and processed to remove nucleases and proteases. The Triple-Pure bead tubes undergo special handling during filling and are quality tested to ensure the elimination of DNase, RNase, protease, and nucleic aci