Learn More
JDM LAB SOLUTIONS LLC Empty 2D bC SBS racks
Supplier:  JDM LAB SOLUTIONS LLC 201501
Encompass
Empty SBS 96 format rack for 0 5ml 2D barcoded storage tubes for use with TPE septum cap only AlteClean
Catalog No. 50-000-14271