Learn More
JDM LAB SOLUTIONS LLC Empty 2D bC SBS racks
Supplier:  JDM LAB SOLUTIONS LLC 201517
Encompass
Empty 196 format rack for all 0 5ml 0 75ml 2D barcoded storage tubes with any cap or thread option AlteClean
Catalog No. 50-000-14217