Learn More
JDM LAB SOLUTIONS LLC Empty 2D bC SBS racks
Supplier:  JDM LAB SOLUTIONS LLC 201503
Encompass
Empty SBS 96 format rack for 0 75ml 2D barcoded storage tubes for use with TPE septum cap only AlteClean
Catalog No. 50-000-14211