Learn More
Genesee Scientific Corporation EMPTY 45ML FPLC COLUMNS
SDP
Catalog No. NC1710741
Encompass
contains:2 columns, 6 frits and 4 caps, 21d x 137h mm, pore size 10um, max pressure(psi/bar/mpa):200/14/1.38
Catalog No. NC1710741 Supplier Genesee Scientific Corporation Supplier No. 20595F45
contains:2 columns, 6 frits and 4 caps, 21d x 137h mm, pore size 10um, max pressure(psi/bar/mpa):200/14/1.38