Learn More
Genesee Scientific Corporation EMPTY 8ML FPLC COLUMNS
SDP
Catalog No. NC1710737
Encompass
contains:3 columns, 9 frits and 6 caps,12dx70h mm, 10um pore size, max pressure(psi/bar/mpa)200/14/1.38
Catalog No. NC1710737 Supplier Genesee Scientific Corporation Supplier No. 20595F8
contains:3 columns, 9 frits and 6 caps,12dx70h mm, 10um pore size, max pressure(psi/bar/mpa)200/14/1.38