Learn More
Montana Molecular Empty Big Sky BacMam negative control - 5mL - Montana Molecular
SDP
Catalog No. NC3787572
Encompass_Preferred
A BacMam vector without any payload - this is a useful tool to examine the effects of BacMam itself on your cells of choice. This uses Montana Molecular's Big Sky BacMam vector. Comes with 500mM sodium butyrate in H2O.
Catalog No. NC3787572 Supplier Montana Molecular Supplier No. X0010N005
A BacMam vector without any payload - this is a useful tool to examine the effects of BacMam itself on your cells of choice. This uses Montana Molecular's Big Sky BacMam vector. Comes with 500mM sodium butyrate in H2O.