Learn More
Molecular Devices Molecular Devices Excitation Filter Slide, Empty, FilterMax F3 F5 Compatible
Catalog No. 5000014577
Encompass_Preferred
The Molecular Devices empty excitation filter slide is an unpopulated slide designed for the installation of user-selected excitation filters in multi-mode microplate readers. It installs into the instrument's filter-slide holder, enabling custom excitation spectra for specialized fluorescence assays.
  • Unpopulated slide for user-installed excitation filters.
  • Allows custom excitation configurations for specific assays.
  • Standard slide form factor for quick installation and removal.
  • Compatible with instruments that accept excitation filter slides per manufacturer specifications.
Catalog No. 50-000-14577 Supplier Molecular Devices Supplier No. 5008587
The Molecular Devices empty excitation filter slide is an unpopulated slide designed for the installation of user-selected excitation filters in multi-mode microplate readers. It installs into the instrument's filter-slide holder, enabling custom excitation spectra for specialized fluorescence assays.
  • Unpopulated slide for user-installed excitation filters.
  • Allows custom excitation configurations for specific assays.
  • Standard slide form factor for quick installation and removal.
  • Compatible with instruments that accept excitation filter slides per manufacturer specifications.