Learn More
Molecular Devices Molecular Devices Filter Slide, Empty, Excitation, 6-Filter Capacity
Catalog No. 5000014418
Encompass_Preferred
Empty excitation filter slide for FilterMax multi-mode microplate readers. Supplied unpopulated to allow custom installation of excitation filters for fluorescence and many luminescence methods. The slide installs into the instrument filter track and secures filters with small hex screws.
  • Supplied empty for custom filter configurations.
  • Accepts up to six individual filters per slide.
  • Distinct sizes for excitation and emission slides to prevent misinstallation.
  • Install and handle by the tab to avoid contaminating filter surfaces.
  • When configured as an excitation slide, not compatible with fluorescence polarization (FP) methods.
Catalog No. 50-000-14418 Supplier Molecular Devices Supplier No. 5008588
Empty excitation filter slide for FilterMax multi-mode microplate readers. Supplied unpopulated to allow custom installation of excitation filters for fluorescence and many luminescence methods. The slide installs into the instrument filter track and secures filters with small hex screws.
  • Supplied empty for custom filter configurations.
  • Accepts up to six individual filters per slide.
  • Distinct sizes for excitation and emission slides to prevent misinstallation.
  • Install and handle by the tab to avoid contaminating filter surfaces.
  • When configured as an excitation slide, not compatible with fluorescence polarization (FP) methods.