Learn More
Molecular Devices Molecular Devices Emission Slide, Empty, Slide 21, FilterMax Compatible
Catalog No. 5000014407
Encompass_Preferred
The empty emission slide is an unpopulated emission filter slide designed to accept user-selected emission filters for custom fluorescence configurations on compatible microplate readers. It installs into the reader's filter slider assembly to provide flexible emission filter selection for fluorescence assays while excluding fluorescence polarization methods.
  • Empty emission filter slide for custom filter installation.
  • Accepts emission filter elements for fluorescence detection.
  • Configurable for fluorescence methods except fluorescence polarization.
  • Supplied as a single slide for in-lab or service installation.
  • Installs into the reader filter slider assembly for filter mounting.
Catalog No. 50-000-14407 Supplier Molecular Devices Supplier No. 5008589
The empty emission slide is an unpopulated emission filter slide designed to accept user-selected emission filters for custom fluorescence configurations on compatible microplate readers. It installs into the reader's filter slider assembly to provide flexible emission filter selection for fluorescence assays while excluding fluorescence polarization methods.
  • Empty emission filter slide for custom filter installation.
  • Accepts emission filter elements for fluorescence detection.
  • Configurable for fluorescence methods except fluorescence polarization.
  • Supplied as a single slide for in-lab or service installation.
  • Installs into the reader filter slider assembly for filter mounting.