Learn More
*AGAROSE BEAD TECHNOLOGIES Empty FPLC 30 mL Column
Catalog No. NC1650472
Encompass_Preferred
FPLC column is a single use product suitable for FPLC and ÄKTA design™ chromatography systems. It is recommended to work with approximately 30ml of packed bead bed volume. The package contains 2 columns, 6 frits (polyethylene frit with a nominal pore size of 12 µm) and 4 caps. The FPLC column parts are made of polypropylene which shows excellent chemical resistance to most of the commonly used reagents
Catalog No. NC1650472 Supplier *AGAROSE BEAD TECHNOLOGIES Supplier No. FPLC302
FPLC column is a single use product suitable for FPLC and ÄKTA design™ chromatography systems. It is recommended to work with approximately 30ml of packed bead bed volume. The package contains 2 columns, 6 frits (polyethylene frit with a nominal pore size of 12 µm) and 4 caps. The FPLC column parts are made of polypropylene which shows excellent chemical resistance to most of the commonly used reagents