Learn More
*AGAROSE BEAD TECHNOLOGIES Empty FPLC 80 ml Columns
Catalog No. NC1650477
Encompass_Preferred
FPLC column is a single use product suitable for FPLC and ÄKTA design™ chromatography systems. It is recommended to work with approximately 80 ml of packed bead bed volume. The package contains 1 column, 3 frits (polyethylene frit with a nominal pore size of 12 µm) and 2 caps. The FPLC column parts are made of polypropylene which shows excellent chemical resistance to most of the commonly used reagents.Tech Specs:Column Material : PolypropyleneFrit Material : PolyethyleneDimensions Diameter x Height (mm) :~ 26 × 144Max Pressure (psi /bar/MPa) :200/14/1.38Frit Pore Size : 10 µmCapacity (ml of packed resin) : 80
Catalog No. NC1650477 Supplier *AGAROSE BEAD TECHNOLOGIES Supplier No. FPLC801
FPLC column is a single use product suitable for FPLC and ÄKTA design™ chromatography systems. It is recommended to work with approximately 80 ml of packed bead bed volume. The package contains 1 column, 3 frits (polyethylene frit with a nominal pore size of 12 µm) and 2 caps. The FPLC column parts are made of polypropylene which shows excellent chemical resistance to most of the commonly used reagents.Tech Specs:Column Material : PolypropyleneFrit Material : PolyethyleneDimensions Diameter x Height (mm) :~ 26 × 144Max Pressure (psi /bar/MPa) :200/14/1.38Frit Pore Size : 10 µmCapacity (ml of packed resin) : 80