Learn More
*AGAROSE BEAD TECHNOLOGIES Empty FPLC 8 mL Columns
Catalog No. NC1650470
Encompass_Preferred
FPLC column is a single use product suitable for FPLC and ÄKTA design™ chromatography systems. It is recommended to work with approximately 8ml of packed bead bed volume. The package contains 3 columns, 9 frits (polyethylene frit with a nominal pore size of 12 µm), and 6 caps. The FPLC column parts are made of polypropylene which shows excellent chemical resistance to most of the commonly used reagents.
Catalog No. NC1650470 Supplier *AGAROSE BEAD TECHNOLOGIES Supplier No. FPLC83
FPLC column is a single use product suitable for FPLC and ÄKTA design™ chromatography systems. It is recommended to work with approximately 8ml of packed bead bed volume. The package contains 3 columns, 9 frits (polyethylene frit with a nominal pore size of 12 µm), and 6 caps. The FPLC column parts are made of polypropylene which shows excellent chemical resistance to most of the commonly used reagents.