Learn More
MilliporeSigma™ Supelco™ Empty Glass Micro-Column (Reversible Tube)
Catalog No. 111009969 Shop All MilliporeSigma Supelco Products
Catalog No. 11-100-9969 Supplier MilliporeSigma™ Supelco™ Supplier No. 28309U
For use with Dioxin Sample Preparation Kit
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Diameter (Metric) Outer 6.35 to 10 mm
Description Pre-Inserted with Stainless Steel Screen to Support Packed Bed
Quantity 10/Pk.
For Use With (Equipment) Dioxin Sample Preparation Kit
Type Micro-Column