Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Empty Optimus Rack for CleanPak and Tip Eject Reloads
GSA_VA

Catalog No. 12111392 Shop All OHAUS Products
Change view
Click to view available options
Size:
10 to 300 μL
1000 to 1250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
12-111-392 1000 to 1250 μL
12-111-391 10 to 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-111-392 Supplier BIOTIX™ Supplier No. 63300152
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BIOTIX™ Empty Optimus Rack for CleanPak and Tip Eject Reloads is compatible with CleanPak and TipEject reloads for easy transfer and racking

  • Locking Clasp to secure tips during transport
  • Full 180 degree hinge design for convenient multichannel pipetting
  • SBS footprint can be used as a 96 well automation reservoir with pyramidal wells
  • Fully autoclaveable for multiple uses with CleanPak and Tip Eject reloads

Order Info

10 racks/pack

Certifications

ISO 9001:2015, Certified free of DNASE, RNASE, Endotoxins/pyrogens, PCR inhibitors. Sterilized product is certified to 10^-3 SAL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description 1000 to 1250μL Optimus Empty Rack
Autoclavable Autoclavable
Color Clear
Holds Pipette Tips (20 to 300 μL, D300)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Application) uTIP and xTIP4 Reloads
For Use With (Equipment) CleanPak and TipEject
Quantity 10 Racks/Pk.
Size 1000 to 1250 μL
Length (Metric) 130 mm
Includes Base and lid
Product Type Empty Refill Rack with Lid
Array 8 x 12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.