Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Empty Pipette Tip Racks for Tips, Nonsterile

Catalog No. 01677343 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1200 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-677-343 10 μL
01-677-203 200 μL
01-677-319 1200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-677-343 Supplier Axygen™ Supplier No. PRFLR300
Add to Cart
Add to Cart

Easy to open hinged lids with a sturdy closing mechanism to keep tips free of contamination.

  • Empty pipette tip racks for Axygen pipette tip reloads, filter pipette tip reloads, and bulk pipette tips.
  • Easy to open hinged lids with a sturdy closing mechanism to keep tips free of contamination.
  • Available for 10 μL, 200 μL, and 1000 μL tips
  • RNase-/DNase-free
  • Human DNA-free
  • Non-Pyrogenic
  • ATP-free
  • Made in facility that complies with current revision of ISO 9001 standard.

Note: This is the suggested replacement for RFL-R.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Gray
Autoclavable Not Autoclavable
Format Reload Rack (tips not included)
Tip Style Standard
Quantity 10 Racks/Pk., 5 Pk/Cs.
Product Type Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.