Learn More
Supply Solutions Sigma Aldrich - Pk20 Empty Fritted 20Ml Spe Tubes; Volume (ML): 20; Porosity (uM): 20; Qty/pk: 20
Catalog No. NC1290839
Encompass_Preferred
Note: PE, PTFE and stainless steel frits for use with polypropylene SPE tubes unless otherwise noted.; Volume (ML): 20; Porosity (uM): 20; Qty/pk: 20
Catalog No. NC1290839 Supplier Supply Solutions Supplier No. 57118U
Note: PE, PTFE and stainless steel frits for use with polypropylene SPE tubes unless otherwise noted.; Volume (ML): 20; Porosity (uM): 20; Qty/pk: 20