Learn More
Supply Solutions Sigma Aldrich - Pk16 Empty Fritted 60Ml Spe Tubes; Volume (ML): 60; Porosity (uM): 20; Qty/pk: 16
Catalog No. NC1290836
Encompass
Note: PE, PTFE and stainless steel frits for use with polypropylene SPE tubes unless otherwise noted.; Volume (ML): 60; Porosity (uM): 20; Qty/pk: 16
Catalog No. NC1290836 Supplier Supply Solutions Supplier No. 57119U
Note: PE, PTFE and stainless steel frits for use with polypropylene SPE tubes unless otherwise noted.; Volume (ML): 60; Porosity (uM): 20; Qty/pk: 16