Learn More
Biotix™ Empty Racks - Removable hinged lid
Catalog No. 12111031 Shop All Biotix Products
Change view
Click to view available options
Holds:
Pipette Tips (20 to 300 μL, D300)
Catalog No. Holds
12-111-031 Pipette Tips (20 to 300 μL, D300)
1 options
This item is not returnable. View return policy
Catalog No. 12-111-031 Supplier Biotix™ Supplier No. M0300TNER
This item is not returnable. View return policy
Features Biotix™ CleanPak™ easily removable reload hinged lid. Biotix™ Empty Racks - Removable hinged lid uses 30-60% less plastic than competitors.
  • Maintains its stability due to its sturdy SBS compliant base
Certifications
ISO 9001 , FDA Class I
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Clear
Holds Pipette Tips (20 to 300 μL, D300)
Material Polypropylene
Positions 96
Shape Rectangular, Rounded Corners
Stackable Yes
Sterility Non-sterile
No. per Case 5 Pks./Cs.
No. per Pack 10 Racks/Pk.
Quantity 960 Tips/Pk., 5 Pks./Cs.
Product Type Empty Rack
Array 8 x 12