Learn More
Axygen™ Empty Tip Rack for 96-well C-Tray Black Conductive Liquid Sensing Automation Tips
Catalog No. 07200813 Shop All Corning Products
Change view
Click to view available options
Holds:
Pipette Tips (1000 μL)
Pipette Tips (50 μL), Pipette Tips (200 μL)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07-200-813 Pipette Tips (1000 μL)
07-200-807 Pipette Tips (50 μL), Pipette Tips (200 μL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-200-813 Supplier Axygen™ Supplier No. TT1000EBATR
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
C-Tray empty tip racks are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • C-tray Format
  • Designed for use with Axygen 96-well C-Tray Black Conductive Liquid Sensing Automation Tips for Tecan Liquid Handling Workstations
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Color Clear/White
Holds Pipette Tips (1000 μL)
Material Polypropylene
Positions 1
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Product Type Empty Rack