Learn More
Hanna Pharmaceutical Supply Co., Inc. Empty INTRAVIA Container 500ML
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 0332B8013
Encompass_Preferred
Empty INTRAVIA Container 500 mL. Sterile, nonpyrogenic fluid path. Port Tubes, which contact fluid path are DEHP-plasticized PVC. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC1648204