Learn More
Epoch Life Science Inc EconoSpin® Empty Columns with inert support for Dye Terminator Removal, Desalting, Pre-filtration etc. With screw cap (50 columns)
SDP
Catalog No. 501538107
Encompass_Preferred
PP housing with 20 um PE frit. 700 ul bed volume. 50-200 ul sample volume. Liquid tight screw cap enables long term buffer storage and eliminates splash when opening/closing. Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 50 columns.
Catalog No. 50-153-8107 Supplier Epoch Life Science Inc Supplier No. 2070050
PP housing with 20 um PE frit. 700 ul bed volume. 50-200 ul sample volume. Liquid tight screw cap enables long term buffer storage and eliminates splash when opening/closing. Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 50 columns.