Learn More
Epoch Life Science Inc EconoSpin® Empty Columns with inert support for Dye Terminator Removal, Desalting, Pre-filtration etc. With attached lid. (50 columns with collection tubes)
SDP
Catalog No. 501538106
Encompass_Preferred
PP housing with 20 um PE frit. 850 ul bed volume. Attached lid simplifies experimental procedure Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 50 columns with collection tubes.
Catalog No. 50-153-8106 Supplier Epoch Life Science Inc Supplier No. 2060050
PP housing with 20 um PE frit. 850 ul bed volume. Attached lid simplifies experimental procedure Ideal for protein desalting or post-sequencingg cleanup for Sanger sequencingg. Ideal for small volume batch mode His-tag protein purification when used with Ni+ agarose. 50 columns with collection tubes.