Learn More
Seca Corp 374 Digital baby scale with extra-large weighing tray
SDP
Catalog No. NC1798766 Shop All Seca Corp Products
Encompass_Preferred
The seca 374 features an extra-large tray with a 44 lbs capacity. The damping feature combined with the auto-HOLD function ensures fast weighing and includes a TARE (net weight) and BMIF (Breast-Milk-Intake-Function). • Weighing tray with high side walls for increased safety • Optional seca 233 length measuring rod • Damping for fast weighing • Suitable for weighing toddlers while sitting • Capacity: 44 lbs / 20 kg • Graduation: 0.2 oz < 22 lbs > 0.5 oz / 5 g < 10 kg > 10 g • Functions: TARE, BMIF, auto-HOLD, auto-CLEAR, lbs/kg switch-over, damping, automatic switch-off • Optional: measuring rod seca 233, head and foot positioners seca 418, power adapter seca 447, power adapter seca 400
Catalog No. NC1798766 Supplier Seca Corp Supplier No. 3741321004
The seca 374 features an extra-large tray with a 44 lbs capacity. The damping feature combined with the auto-HOLD function ensures fast weighing and includes a TARE (net weight) and BMIF (Breast-Milk-Intake-Function). • Weighing tray with high side walls for increased safety • Optional seca 233 length measuring rod • Damping for fast weighing • Suitable for weighing toddlers while sitting • Capacity: 44 lbs / 20 kg • Graduation: 0.2 oz < 22 lbs > 0.5 oz / 5 g < 10 kg > 10 g • Functions: TARE, BMIF, auto-HOLD, auto-CLEAR, lbs/kg switch-over, damping, automatic switch-off • Optional: measuring rod seca 233, head and foot positioners seca 418, power adapter seca 447, power adapter seca 400