Learn More
Seca Corp 664 EMR-validated digital wheelchair scale
SDP
Catalog No. NC1798771 Shop All Seca Corp Products
Encompass_Preferred
With its high capacity of 800 pounds, seca 664 wheelchair scale makes it easy to weigh all patient types. Easy access is assured by its extremely low platform ramps. The secure side rails give additional safety. The swiveling LCD with large letters is easy-to-read both for the patient and for the caregiver. The seca 664 can be folded up in no time and can be transported anywhere, thanks to its roller castors. Measured data can be wirelessly transmitted to an EMR system. • Side rails on the platform ensure safe rolling of a wheelchair on/off scale platform • High capacity of up to 800 pounds • Flat platform with self-lowering ramps • Two ramps included • Easy to fold together and transport • EMR-validated: transmit measured data directly to any EMR system • Capacity: 800 lbs / 360 kg • Graduation: 0.1 lbs / 50 g • Functions: TARE, pre-TARE, HOLD, auto-HOLD, BMI, auto-CLEAR, lbs/kg switch-over, damping, automatic switch-off
Catalog No. NC1798771 Supplier Seca Corp Supplier No. 6641321103
With its high capacity of 800 pounds, seca 664 wheelchair scale makes it easy to weigh all patient types. Easy access is assured by its extremely low platform ramps. The secure side rails give additional safety. The swiveling LCD with large letters is easy-to-read both for the patient and for the caregiver. The seca 664 can be folded up in no time and can be transported anywhere, thanks to its roller castors. Measured data can be wirelessly transmitted to an EMR system. • Side rails on the platform ensure safe rolling of a wheelchair on/off scale platform • High capacity of up to 800 pounds • Flat platform with self-lowering ramps • Two ramps included • Easy to fold together and transport • EMR-validated: transmit measured data directly to any EMR system • Capacity: 800 lbs / 360 kg • Graduation: 0.1 lbs / 50 g • Functions: TARE, pre-TARE, HOLD, auto-HOLD, BMI, auto-CLEAR, lbs/kg switch-over, damping, automatic switch-off