Learn More
Seca Corp 676 EMR-validated wheelchair scale with robust handrail and transport wheels
SDP
Catalog No. NC1798772 Shop All Seca Corp Products
Encompass_Preferred
Thanks to its large platform and sturdy handrail, the seca 676 is extremely versatile. Patients can be weighed while standing or sitting in a wheelchair or chair. The handrail provides support for patients when needed. It's high capacity makes the scale ideal for weighing heavy patients. The handrail also serves as handle when the scale is folded, allowing the scale to be moved around or stowed away effortlessly and quickly. • Large wheelchair-accessible platform • Convenient and foldable to save space • Mobile with transport wheels • Handrail as a standing aid • Non-slip rubber mat • Two ramps included • EMR-validated: transmit measured data directly to EMR system • Capacity: 800 lbs / 360 kg • Graduation: 0.1 lbs / 50 g • Functions: TARE, pre-TARE, HOLD, auto-HOLD, BMI, auto-CLEAR, lbs/kg switch-over, damping
Catalog No. NC1798772 Supplier Seca Corp Supplier No. SECA 676
Thanks to its large platform and sturdy handrail, the seca 676 is extremely versatile. Patients can be weighed while standing or sitting in a wheelchair or chair. The handrail provides support for patients when needed. It's high capacity makes the scale ideal for weighing heavy patients. The handrail also serves as handle when the scale is folded, allowing the scale to be moved around or stowed away effortlessly and quickly. • Large wheelchair-accessible platform • Convenient and foldable to save space • Mobile with transport wheels • Handrail as a standing aid • Non-slip rubber mat • Two ramps included • EMR-validated: transmit measured data directly to EMR system • Capacity: 800 lbs / 360 kg • Graduation: 0.1 lbs / 50 g • Functions: TARE, pre-TARE, HOLD, auto-HOLD, BMI, auto-CLEAR, lbs/kg switch-over, damping