Learn More
Seca Corp 787 EMR-validated column scale with eye-level display, handrail-compatible
SDP
Catalog No. NC1798785 Shop All Seca Corp Products
Encompass_Preferred
The seca 787 EMR-validated column scale with eye-level display sends results directly into your EMR. The digital measuring rod has an extra-long measuring range (4 – 90 inches).With an upgradable network module (seca 452), the seca 787 transmits measurements directly to EMR system. • Medical-grade, robust scale for years of use with high patient volume • Digital measuring rod with an extra long measuring range • 8-year warranty included (4x industry standard) • Two-Line display shows height and weight • Damping system and Auto-HOLD guarantee quick and Precise Weighing • Oversized, low-profile full metal platform (no plastic) • Tilt-proof safety design • Capacity 550 lbs / 250 kg • Graduation 0.2 lb / 100 g
Catalog No. NC1798785 Supplier Seca Corp Supplier No. 7871321004
The seca 787 EMR-validated column scale with eye-level display sends results directly into your EMR. The digital measuring rod has an extra-long measuring range (4 – 90 inches).With an upgradable network module (seca 452), the seca 787 transmits measurements directly to EMR system. • Medical-grade, robust scale for years of use with high patient volume • Digital measuring rod with an extra long measuring range • 8-year warranty included (4x industry standard) • Two-Line display shows height and weight • Damping system and Auto-HOLD guarantee quick and Precise Weighing • Oversized, low-profile full metal platform (no plastic) • Tilt-proof safety design • Capacity 550 lbs / 250 kg • Graduation 0.2 lb / 100 g