Learn More
Aviva Systems Biology Corp ADGRE1 Antibody - C-terminal region (ARP64411_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64411_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EMR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human EMR1. Alternate names: TM7LN3ApplicationWB. Species reactivity: Human
Catalog No. 50-288-257