Learn More
Medchemexpress LLC HY-10396 10mg Medchemexpress, Emricasan CAS:254750-02-2 Purity:>98%
SDP
Catalog No. 501968178
Encompass_Preferred
Medchemexpress, HY-10396 10mg Emricasan CAS:254750-02-2 Emricasan (PF 03491390; IDN-6556) is an irreversible pan-caspase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8178 Supplier Medchemexpress LLC Supplier No. HY1039610MG
Medchemexpress, HY-10396 10mg Emricasan CAS:254750-02-2 Emricasan (PF 03491390; IDN-6556) is an irreversible pan-caspase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.