Learn More
Medchemexpress LLC HY-10396 50mg , Emricasan CAS:254750-02-2 Purity:>98%
SDP
Catalog No. 502259121
Encompass_Preferred
HY-10396 50mg Emricasan CAS: 254750-02-2 Emricasan (PF 03491390) is an orally active and irreversible pan-caspase inhibitor. Emricasan inhibits Zika virus (ZIKV)-induced increases in caspase-3 activity and protected human cortical neural progenitors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9121 Supplier Medchemexpress LLC Supplier No. HY1039650MG
HY-10396 50mg Emricasan CAS: 254750-02-2 Emricasan (PF 03491390) is an orally active and irreversible pan-caspase inhibitor. Emricasan inhibits Zika virus (ZIKV)-induced increases in caspase-3 activity and protected human cortical neural progenitors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.