Learn More
Electron Microscopy Sciences EMS 100 Grid Storage Box
SDP
Catalog No. 5034562
Encompass_Preferred
Used for the storage of TEM grids for routine grid handling, transport and long term TEM grid storage. For standard grids that are 3.05 mm in diameter. Anti Static treated
Catalog No. 50-345-62 Supplier Electron Microscopy Sciences Supplier No. 7114602
Used for the storage of TEM grids for routine grid handling, transport and long term TEM grid storage. For standard grids that are 3.05 mm in diameter. Anti Static treated