Learn More
Electron Microscopy Sciences Microscope Immersion Oils, Refractive index: 1.518 ± 0.002., 1/EA
SDP
Catalog No. 502090559
Encompass_Preferred
EMS is proud to introduce a complete new line of immersion oils. EMS 250 is a medium viscosity oil formed from long chain hydrocarbons and polybutenes and terphenyls with a R.I of 1.518 ± 0.002 with water-white to a light straw color. Refractive index: 1.518 ± 0.002.
Catalog No. 50-209-0559 Supplier Electron Microscopy Sciences Supplier No. 1600501
EMS is proud to introduce a complete new line of immersion oils. EMS 250 is a medium viscosity oil formed from long chain hydrocarbons and polybutenes and terphenyls with a R.I of 1.518 ± 0.002 with water-white to a light straw color. Refractive index: 1.518 ± 0.002.