Learn More
Electron Microscopy Sciences Microscope Immersion Oils, Refractive index: 1.480., 1/EA
SDP
Catalog No. 502090563
Encompass_Preferred
EMS is proud to introduce a complete new line of immersion oils. EMS 265 is a low viscosity immersion oil with a R.I of 1.480. Refractive index: 1.480.
Catalog No. 50-209-0563 Supplier Electron Microscopy Sciences Supplier No. 1600604
EMS is proud to introduce a complete new line of immersion oils. EMS 265 is a low viscosity immersion oil with a R.I of 1.480. Refractive index: 1.480.