Learn More
Electron Microscopy Sciences EMS 3100 Service Kit
SDP
Catalog No. NC3450751
Encompass
E3100-1 Specimen holder for 3.05 mm grids, E3100-2 Specimen holder for 2.3 mm grids E3100-01 Specimen holder for tissue (boat), E3100-02 Specimen holder for coverslips, EMS 800A Porous pots with lids 12.7 mm x 15.5 mm (pack of 10) for micro-specimens E3500 Thermocirculator for control of heating cyclem E4860 Recirculating Heater/Chiller to control heating and cooling cycle (please specify voltage)
Catalog No. NC3450751 Supplier Electron Microscopy Sciences Supplier No. E3100061
E3100-1 Specimen holder for 3.05 mm grids, E3100-2 Specimen holder for 2.3 mm grids E3100-01 Specimen holder for tissue (boat), E3100-02 Specimen holder for coverslips, EMS 800A Porous pots with lids 12.7 mm x 15.5 mm (pack of 10) for micro-specimens E3500 Thermocirculator for control of heating cyclem E4860 Recirculating Heater/Chiller to control heating and cooling cycle (please specify voltage)