Learn More
Electron Microscopy Sciences EMS Oscillating Tissue Slicer Model 4500
SDP
Catalog No. 5025768
Encompass_Preferred
The EMS 4500 and 5000 Oscillating Tissue Slicer are for sectioning either fixed or fresh tissue without embedding or freezing your sample. Preparation time is reduced dramatically and the risk of distortion and artifacts associated with these procedures are eliminated. The EMS 4500 and 5000 meets the highest standards for precision and accuracy and exceeds the performance of units on the market costing twice as much.
Catalog No. 50-257-68 Supplier Electron Microscopy Sciences Supplier No. 93200
Add to Cart
Add to Cart
Request a Quote
The EMS 4500 and 5000 Oscillating Tissue Slicer are for sectioning either fixed or fresh tissue without embedding or freezing your sample. Preparation time is reduced dramatically and the risk of distortion and artifacts associated with these procedures are eliminated. The EMS 4500 and 5000 meets the highest standards for precision and accuracy and exceeds the performance of units on the market costing twice as much.