Learn More
Electron Microscopy Sciences EMS Aluminium Micro-Punch Kit, 1.2 mm
SDP
Catalog No. NC1508075
Encompass_Preferred
The EMS Aluminum Punch Kit is designed to serve research scientists with a dissection tool for small, pre-defined regions of the tissue or the specimen. Barrel is machined from high grade 6061 light weight, seamless, aluminium tubing and anodized to form a protective, chemically resistant surface. Easy handling. All punches are 130 mm in length, with a shaft of 3.2 mm and a tip length of 16.8 mm. Deluxe plastic case included.
Size: 1.2 mm
Catalog No. NC1508075 Supplier Electron Microscopy Sciences Supplier No. 6903312
The EMS Aluminum Punch Kit is designed to serve research scientists with a dissection tool for small, pre-defined regions of the tissue or the specimen. Barrel is machined from high grade 6061 light weight, seamless, aluminium tubing and anodized to form a protective, chemically resistant surface. Easy handling. All punches are 130 mm in length, with a shaft of 3.2 mm and a tip length of 16.8 mm. Deluxe plastic case included.
Size: 1.2 mm