Learn More
Electron Microscopy Sciences Contoured Isolation Platform AMC-7 Platform, White Epoxy, 580 x 380 mm x 45 mm
SDP
Catalog No. 501902643
Encompass_Preferred
Made from high grade steel parts welded together for rigidity and coated with a tough epoxy powder white finish. Three elastomeric isolators remove over 97% of horizontal vibrations at 50Hz and over 95% of vertical vibrations.
Size: White Epoxy
Surface: 580 x 380 mm x 45 mm
Catalog No. 50-190-2643 Supplier Electron Microscopy Sciences Supplier No. 6719
Made from high grade steel parts welded together for rigidity and coated with a tough epoxy powder white finish. Three elastomeric isolators remove over 97% of horizontal vibrations at 50Hz and over 95% of vertical vibrations.
Size: White Epoxy
Surface: 580 x 380 mm x 45 mm