Learn More
Electron Microscopy Sciences EMS Autopsy Grossing Knife, 8" blade
SDP
Catalog No. 501931605
Encompass_Preferred
Our Sani-Safe autopsy grossing knives have plastic non-slip safety handles that comfortably mold to your hand to reduce fatigue, as well as slip guards to protect your hands from the cutting edge. The handle is securely bonded to the cutting blade. The blade is well-balanced, hand-ground and honed, making it the perfect autopsy knife. It can be constantly sharpened for long life and easily sanitized with any disinfectant. These knives are made with premium grade stainless steel.
Blade: 8"
Catalog No. 50-193-1605 Supplier Electron Microscopy Sciences Supplier No. 7206003
Add to Cart
Add to Cart
Our Sani-Safe autopsy grossing knives have plastic non-slip safety handles that comfortably mold to your hand to reduce fatigue, as well as slip guards to protect your hands from the cutting edge. The handle is securely bonded to the cutting blade. The blade is well-balanced, hand-ground and honed, making it the perfect autopsy knife. It can be constantly sharpened for long life and easily sanitized with any disinfectant. These knives are made with premium grade stainless steel.
Blade: 8"