Learn More
Electron Microscopy Sciences EMS Bell Jar Shine 1 LB
SDP
Catalog No. 501901833
Encompass_Preferred
This unique cleaning compound will not only clean but it polishes as well. Meant to be used on any surface which is either glass, metal, porcelain, ceramic and even chrome surfaces. Perfect for the cleaning of your bell jars from your Carbon and Sputter Coaters, as well as any surfaces in the lab that are metal, ceramic, steel etc.
Catalog No. 50-190-1833 Supplier Electron Microscopy Sciences Supplier No. 6054001
This unique cleaning compound will not only clean but it polishes as well. Meant to be used on any surface which is either glass, metal, porcelain, ceramic and even chrome surfaces. Perfect for the cleaning of your bell jars from your Carbon and Sputter Coaters, as well as any surfaces in the lab that are metal, ceramic, steel etc.