Learn More
Electron Microscopy Sciences Cabinet System Base
SDP
Catalog No. 5028757
Encompass_Preferred
Our newly introduced cabinets for filing 3x1" slides is the most effective system available to the ever-growing problem of slide storage and retrieval. They are compact, sturdy and economical. The cabinets are stackable and are secured by interlocks. Store up to 54,000 slides in only 2 square feet of space. Low cost slide storage. Ideal for use in Pathology, Cytology, Hematology, Histology, Dermatology, Research, Education. Base lifts cabinet 4 3/4".
Catalog No. 50-287-57 Supplier Electron Microscopy Sciences Supplier No. 7478010
Our newly introduced cabinets for filing 3x1" slides is the most effective system available to the ever-growing problem of slide storage and retrieval. They are compact, sturdy and economical. The cabinets are stackable and are secured by interlocks. Store up to 54,000 slides in only 2 square feet of space. Low cost slide storage. Ideal for use in Pathology, Cytology, Hematology, Histology, Dermatology, Research, Education. Base lifts cabinet 4 3/4".