Learn More
Electron Microscopy Sciences Cured Epoxy Remover
SDP
Catalog No. 50980449
Encompass_Preferred
Studies show that in order to get good results in positive staining of sections, the removal of cured epoxy (embedding material) as well as the reduced-osmium (fixation materials) from the sections is necessary; especially when double fixation with an Aldehyde and Osmium tetroxide is employed to process specimens for E.M. We offer a CURED-EPOXY-REMOVER kit, which removes the polymerized resin as well as the reduced-osmium. The structure of the resin free section will remain unchanged.
Catalog No. 50-980-449 Supplier Electron Microscopy Sciences Supplier No. 14942
Studies show that in order to get good results in positive staining of sections, the removal of cured epoxy (embedding material) as well as the reduced-osmium (fixation materials) from the sections is necessary; especially when double fixation with an Aldehyde and Osmium tetroxide is employed to process specimens for E.M. We offer a CURED-EPOXY-REMOVER kit, which removes the polymerized resin as well as the reduced-osmium. The structure of the resin free section will remain unchanged.