Learn More
Electron Microscopy Sciences EMS Disc Punch Set
SDP
Catalog No. 50980920
Encompass
Prepare high-quality disk specimens from thin metal foils with the EMS 130 Specimen Punch. A precision ground punch and die plate eliminate specimen stress and distortion. For convenient handling, a spring-loaded return plunger keeps the disk specimen on the die plate surface.
: Wide base
: 4.0" x 4.0"
: 3.1"
Catalog No. 50-980-920 Supplier Electron Microscopy Sciences Supplier No. 5012510
Prepare high-quality disk specimens from thin metal foils with the EMS 130 Specimen Punch. A precision ground punch and die plate eliminate specimen stress and distortion. For convenient handling, a spring-loaded return plunger keeps the disk specimen on the die plate surface.
: Wide base
: 4.0" x 4.0"
: 3.1"