Learn More
Electron Microscopy Sciences EMS Economy Tweezers with Serrations Style 3C, 110 mm OAL
SDP
Catalog No. 501930925
Encompass_Preferred
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 110 mm
OAL: Straight, very sharp, fine
Catalog No. 50-193-0925 Supplier Electron Microscopy Sciences Supplier No. 7832645
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 110 mm
OAL: Straight, very sharp, fine