Learn More
Electron Microscopy Sciences EMS Economy Tweezers with Serrations Style 7A, 120 mm OAL
SDP
Catalog No. 501930931
Encompass_Preferred
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 120 mm
OAL: Fine, curved
Catalog No. 50-193-0931 Supplier Electron Microscopy Sciences Supplier No. 7832651
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 120 mm
OAL: Fine, curved