Learn More
Electron Microscopy Sciences EMS Economy Tweezers with Serrations Style AC, 110 mm OAL
SDP
Catalog No. 501930933
Encompass_Preferred
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 110 mm
OAL: Straight, strong
Catalog No. 50-193-0933 Supplier Electron Microscopy Sciences Supplier No. 7832653
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 110 mm
OAL: Straight, strong