Learn More
Electron Microscopy Sciences EMS Economy Tweezers with Serrations Style 3, 120 mm OAL
SDP
Catalog No. 501930924
Encompass_Preferred
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 120 mm
OAL: Straight, very sharp, fine
Catalog No. 50-193-0924 Supplier Electron Microscopy Sciences Supplier No. 7832644
The tweezers are made from 304 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 120 mm
OAL: Straight, very sharp, fine