Learn More
Electron Microscopy Sciences EMS 9002 Vacuum Processor
SDP
Catalog No. 5029051
Encompass_Preferred
Made from Pyrex Glass, which offers better temperature conductivity and stability than any other material and is chemically resistant and microwave transparent. The EMS 9002 when used in conjunction with our EMS 9000 processor improves ultra structural preservation during microwave assisted chemical fixation and reduces fixation and infiltration times dramatically.
Catalog No. 50-290-51 Supplier Electron Microscopy Sciences Supplier No. 97050A
Made from Pyrex Glass, which offers better temperature conductivity and stability than any other material and is chemically resistant and microwave transparent. The EMS 9002 when used in conjunction with our EMS 9000 processor improves ultra structural preservation during microwave assisted chemical fixation and reduces fixation and infiltration times dramatically.